2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Sodium 2'-(dicyclohexylphosphino)-2,6-dimethoxy-[1,1'-biphenyl]-3-sulphonate hydrate, (water soluble SPhos)

A175758-1G A175758-25G A175758-250MG A175758-100G
103807-086EA 30 USD
103807-086 103845-398 77082-742 77082-744 77593-116 77683-412
2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt 98%, Ambeed.Inc
2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt

Precaución: For research use only.

Formula: C₂₆H₃₆NaO₆PS
Storage Temperature: Refrigerator
MDL Number: MFCD07781994
CAS Number: 1049726-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR