Loxoprofen sodium 97%

Supplier: Ambeed
Warning

A100810-100G A100810-1G A100810-250MG A100810-25G A100810-10G A100810-1MG A100810-500G
103821-332EA 15 USD
103821-332 76869-108 76869-110 76869-112 76869-114 77670-454 77670-456 77670-458
Loxoprofen sodium 97%
Loxoprofen sodium

Precaución: for Research Use Only

Formula: C₁₅H₁₇NaO₃
Boiling Pt: 417.9 °C (760 mm Hg)
Flash Pt: 220.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00941425
CAS Number: 80382-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR