1-Pentanesulfonic acid sodium salt monohydrate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium 1-pentanesulphonate monohydrate
A706978-25G
A706978-5G
A706978-100G
103840-676EA
20
USD
103840-676
76844-890
76844-892
77243-540
1-Pentanesulfonic acid sodium salt monohydrate 99%
1-Pentanesulphonic acid sodium salt monohydrate
Precaución: for Research Use Only
Formula:
C₅H₁₁NaO₃S·1H₂O MW: 192.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 207605-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...