Diethyl oxalacetate sodium salt 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium diethyl oxalacetate, Sodium Diethyl Oxalacetate, Oxalacetic Acid Diethyl Ester Sodium Salt
A247452-25G
A247452-100G
A247452-10G
A247452-500G
A247452-1000G
103809-762EA
181
USD
103809-762
103851-604
77090-944
77113-374
77113-376
77113-378
77304-406
Diethyl oxalacetate sodium salt 95%
Diethyl oxalacetate sodium salt
Precaución: for Research Use Only
Formula:
C₈H₁₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00035571 CAS Number: 40876-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...