ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) 98%

Supplier: Ambeed
Warning

A341469-1MG A341469-1G A341469-100MG A341469-250MG A341469-5G
103799-614EA 28 USD
103799-614 103825-836 76693-976 77306-538 77550-982 77550-984 77626-648
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) 98%
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

Precaución: for Research Use Only

Formula: C₂₁H₂₆N₇Na₄O₁₇P₃
Boiling Pt: 1175.1 °C (760 mm Hg)
Melting Pt: >250 °C
Flash Pt: 664.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00036263
CAS Number: 2646-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR