L-Cystine disodium salt 99%

Supplier: Ambeed
Warning

Synonyms: Sodium (2R,2'R)-3,3'-disulphanediylbis(2-aminopropanoate)

A216491-100G A216491-10G A216491-500G A216491-25G A216491-5G
103809-518EA 202 USD
103809-518 103849-282 76800-776 76800-778 76800-780 77087-870 77087-872
L-Cystine disodium salt 99%
L-Cystine disodium salt

Precaución: for Research Use Only

Formula: C₆H₁₀N₂Na₂O₄S₂
MW: 284.30 g/mol
Melting Pt: 220 °C
Flash Pt: 237 °C
Storage Temperature: Ambient
MDL Number: MFCD00037206
CAS Number: 64704-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR