SN-glycero-3-phosphocholine 98%

Supplier: Ambeed
Warning

A169170-10G A169170-250MG A169170-1G A169170-25G A169170-5G A169170-500G A169170-100G
103806-360EA 7 USD
103806-360 103844-614 76799-334 76799-336 77081-682 77081-684 77081-686 77593-046 77600-360
SN-glycero-3-phosphocholine 98%
SN-glycero-3-phosphocholine

Precaución: for Research Use Only

Formula: C₈H₂₀NO₆P
Storage Temperature: Freezer
MDL Number: MFCD00063544
CAS Number: 28319-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR