sec-Butyl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

A227015-1G A227015-5G A227015-10G A227015-250MG
103819-288EA 39 USD
103819-288 103850-196 77088-946 77088-948 77165-420 77165-422
sec-Butyl 4-methylbenzenesulfonate 95%
sec-Butyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃S
MW: 228.3 g/mol
Boiling Pt: 329.1 °C (760 mm Hg)
Flash Pt: 152.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038770
CAS Number: 715-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR