sec-Butyl 2-(2-hydroxyethyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Icaridin

A203941-100G A203941-10G A203941-500G A203941-25G A203941-1G A203941-5G
103817-510EA 5 USD
103817-510 103848-170 76800-448 76800-450 76800-452 77086-316 77302-886 77302-888
sec-Butyl 2-(2-hydroxyethyl)piperidine-1-carboxylate 98%
sec-Butyl 2-(2-hydroxyethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₃
MW: 229.31 g/mol
Boiling Pt: 330.9 °C (760 mm Hg)
Flash Pt: 153.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01756488
CAS Number: 119515-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR