Scandium (III) trifluoromethanesulfonate 99%

Supplier: Ambeed
Danger

Synonyms: Scandium(III) trifluoromethanesulphonate, Scandium triflate

A102372-100G A102372-250MG A102372-1G
103796-364EA 14 USD
103796-364 103822-058 77148-282 77148-284 77148-286 77148-288 77308-944 77308-946
Scandium (III) trifluoromethanesulfonate 99%
Scandium(III) trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₃F₉O₉S₃Sc
MW: 492.17 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00192433
CAS Number: 144026-79-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR