SB 225002 98%

Supplier: Ambeed
Warning

Synonyms: SB225002, 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea

A149514-10MG A149514-1MG A149514-25MG A149514-50MG A149514-5MG A149514-100MG A149514-250MG
103804-052EA 252 USD
103804-052 103842-168 77190-738 77190-740 77190-742 77190-744 77190-746 77615-206 77615-208
SB 225002 98%
SB 225002

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrN₃O₄
MW: 352.10 g/mol
Boiling Pt: 401.6 °C (760 mm Hg)
Flash Pt: 196.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00954637
CAS Number: 182498-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR