SB203580 98%

Supplier: Ambeed
Warning

Synonyms: SB 203580, 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine

A248150-1MG A248150-5MG A248150-100MG A248150-250MG
103791-590EA 29 USD
103791-590 103792-492 103799-526 103811-814 76722-568 77304-440 77304-442 77668-024 77668-026
SB203580 98%
SB203580

Precaución: for Research Use Only

Formula: C₂₁H₁₆FN₃OS
MW: 377.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00922198
CAS Number: 152121-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR