2',3'-Didehydro-3'-deoxythymidine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Stavudine, 2',3'-Anhydrothymidine, 3'-Deoxy-2',3'-didehydrothymidine, 1-(2,3-Dideoxy-beta-D-glycero-2-pentenofuranosyl)thymine, 1-((2R,5S)-5-(Hydroxymethyl)-2,5-dihydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, d4T, Sanilvudine
A317604-100MG
A317604-10G
A317604-1G
A317604-100G
103825-140EA
16
USD
103825-140
103836-070
76788-002
76788-004
76838-922
77668-406
2',3'-Didehydro-3'-deoxythymidine 95%
2',3'-Didehydro-3'-deoxythymidine
Precaución: for Research Use Only
Formula:
C₁₀H₁₂N₂O₄ MW: 224.22 g/mol Melting Pt: 159…160 °C Storage Temperature: Freezer |
MDL Number:
MFCD00132921 CAS Number: 3056-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...