(S)-2-Pyridylthio cysteamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridyldithio)ethylamine HCl, S-(2-Pyridylthio)cysteamine hydrochloride

A288383-100MG A288383-250MG A288383-1G A288383-5G A288383-10G A288383-25G
103813-794EA 140 USD
103813-794 103834-936 76723-432 76723-434 77305-530 77305-532 77571-788 77668-280
(S)-2-Pyridylthio cysteamine hydrochloride 97%
(S)-2-Pyridylthio cysteamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀N₂S₂·ClH
MW: 222.759 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD01863453
CAS Number: 106139-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR