(S)-(+)-Mandelic acid 98%

Supplier: Ambeed
Warning

Synonyms: L(+)-Mandelic acid, (S)-(+)-2-Hydroxy-2-phenylacetic acid, L(+)-2-Hydroxy-2-phenylacetic acid, (S)-2-Hydroxy-2-phenylacetic acid

A218130-100G A218130-10G A218130-1G A218130-500G A218130-25G A218130-1000G
103809-532EA 57 USD
103809-532 76691-700 76691-702 76691-704 76691-706 77088-080 77303-480
(S)-(+)-Mandelic acid 98%
(S)-(+)-Mandelic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 321.8 °C (760 mm Hg)
Melting Pt: 131…134 °C
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004495
CAS Number: 17199-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR