Ruthenium(IV) oxide hydrate ≥99%

Supplier: Ambeed
Warning

A782289-10G A782289-250MG
103840-986EA 138 USD
103840-986 77873-243 77873-244
Ruthenium(IV) oxide hydrate ≥99%
Ruthenium(IV) oxide hydrate

Precaución: Inert atmosphere, room temperature

Formula: O₂Ru·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149846
CAS Number: 32740-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR