Ruthenium(III) chloride hydrate 97%

Supplier: Ambeed
Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

A708346-10G A708346-1G A708346-25G A708346-5G
103840-688EA 142 USD
103840-688 76831-684 76844-904 77308-176 77308-178
Ruthenium(III) chloride hydrate 97%
Ruthenium(III) chloride hydrate

Precaución: for Research Use Only

Formula: Cl₃Ru·xH₂O
MW: 207.43 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00149844
CAS Number: 14898-67-0
UN: 3260
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR