(R)-Roscovitine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine, Roscovitine, (R)-2-((6-(Benzylamino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol
A300507-1MG
A300507-5MG
A300507-10MG
103792-576EA
71
USD
103792-576
103799-582
103814-274
76723-618
76723-620
76838-108
(R)-Roscovitine 98%
(R)-Roscovitine
Precaución: for Research Use Only
Formula:
C₁₉H₂₆N₆O MW: 354.45 g/mol Melting Pt: 106…107 °C Storage Temperature: Freezer |
MDL Number:
MFCD02266401 CAS Number: 186692-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...