Roflumilast ≥99%

Supplier: Ambeed
Warning

A168666-10MG A168666-50MG A168666-5MG A168666-1G A168666-5G A168666-250MG A168666-100G
103806-310EA 33 USD
103806-310 103844-564 76720-402 76720-404 76720-406 77081-622 77081-624 77667-354 77871-846
Roflumilast ≥99%
Roflumilast

Precaución: for Research Use Only

Formula: C₁₇H₁₄Cl₂F₂N₂O₃
Boiling Pt: 430.6 °C (760 mm Hg)
Melting Pt: 158 °C
Storage Temperature: Freezer
MDL Number: MFCD00938270
CAS Number: 162401-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR