Rivastigmine 98%

Supplier: Ambeed
Warning

Synonyms: Exelon, SDZ-ENA 713, S-Rivastigmine, (S)-3-(1-(Dimethylamino)ethyl)phenyl ethyl(methyl)carbamate

A248924-1G A248924-100MG A248924-50MG A248924-250MG
103811-848EA 130 USD
103811-848 103851-682 77304-466 77600-998 77601-000 77668-030
Rivastigmine 98%
Rivastigmine

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₂
MW: 250.34 g/mol
Boiling Pt: 316.2 °C (760 mm Hg)
Flash Pt: 145 °C
Storage Temperature: Freezer
MDL Number: MFCD00871496
CAS Number: 123441-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR