Rhodium(III) chloride hydrate 98%

Supplier: Ambeed
Warning

A885429-250MG A885429-1G A885429-100MG
103841-324EA 2974 USD
103841-324 76729-030 76845-592 77603-220
Rhodium(III) chloride hydrate 98%
Rhodium(III) chloride hydrate

Precaución: for Research Use Only

Formula: Cl₃Rh·H₂O
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00149839
CAS Number: 20765-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR