Rhodium(III) chloride trihydrate 98%

Supplier: Ambeed
Warning

A294104-100MG A294104-250MG A294104-1G
103813-998EA 656 USD
103813-998 76723-522 76723-524 76837-764
Rhodium(III) chloride trihydrate 98%
Rhodium(III) chloride trihydrate

Precaución: for Research Use Only

Formula: Cl₃Rh·3H₂O
MW: 263.31 g/mol
Boiling Pt: 800 °C (760 mm Hg)
Melting Pt: 475 °C
Storage Temperature: Ambient
MDL Number: MFCD00149840
CAS Number: 13569-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR