Retinoic acid 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
A126336-100MG
A126336-10G
A126336-1G
A126336-25G
A126336-5G
A126336-500G
A126336-100G
103800-796EA
12
USD
103800-796
77285-124
77285-126
77298-112
77298-114
77298-116
77571-564
77614-986
Retinoic acid 98%
Retinoic acid
Precaución: for Research Use Only
Formula:
C₂₀H₂₈O₂ MW: 300.44 g/mol Boiling Pt: 462.8 °C (760 mm Hg) Melting Pt: 179…184 °C Flash Pt: 350.6 °C Storage Temperature: Freezer |
MDL Number:
MFCD00001551 CAS Number: 302-79-4 UN: 3077 ADR: 9,Ⅲ |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...