trans-tert-Butyl-6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate 95%

Supplier: Ambeed
Warning

A193849-100MG A193849-250MG A193849-25G A193849-1G A193849-5G
103816-590EA 78 USD
103816-590 103847-198 76974-944 76974-946 76974-948 77302-366 77302-368
trans-tert-Butyl-6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate 95%
trans-tert-Butyl-6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.27 g/mol
Boiling Pt: 275.7±33.0 °C (760 mm Hg)
Flash Pt: 120.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15111971
CAS Number: 273206-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR