tert-Butyl (1R,5S,6R)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate 95%

Supplier: Ambeed
Warning

A254563-250MG A254563-1G
103812-144EA 62 USD
103812-144 103851-998 77014-066 77304-634
tert-Butyl (1R,5S,6R)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate 95%
tert-Butyl (1R,5S,6R)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
Boiling Pt: 307.5±15.0 °C (760 mm Hg)
Flash Pt: 139.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14525755
CAS Number: 419572-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR