Rapamycin ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
A656002-1G
A656002-25MG
A656002-100MG
A656002-250MG
A656002-5G
103799-710EA
15
USD
103799-710
103840-416
77573-106
77573-108
77594-996
77614-024
77614-026
Rapamycin ≥98%
Rapamycin
Precaución: Sealed in dry, store in freezer, under –20 °C.
Formula:
C₅₁H₇₉NO₁₃ MW: 914.19 g/mol Boiling Pt: 973 °C (760 mm Hg) Flash Pt: 542.3 °C Storage Temperature: Freezer |
MDL Number:
MFCD00867594 CAS Number: 53123-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...