racemic-4,12-Dibromo[2.2]paracyclophane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Racemic-4,12-dibromo[2.2]paracyclophane
A139970-250MG
A139970-1G
A139970-5G
A139970-25G
103802-806EA
27
USD
103802-806
77162-262
77315-914
77315-916
77683-222
racemic-4,12-Dibromo[2.2]paracyclophane 97%
racemic-4,12-Dibromo[2.2]paracyclophane
Precaución: For research use only.
Formula:
C₁₆H₁₄Br₂ MW: 366.0904 g/mol Melting Pt: 250…251 °C Storage Temperature: Ambient |
MDL Number:
MFCD00184346 CAS Number: 23927-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...