Racecadotril 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl N-[3-(Acetylthio)-2-benzylpropionyl]glycinate, Acetorphan, N-[3-(Acetylthio)-2-benzylpropionyl]glycine Benzyl Ester

A148453-100G A148453-10G A148453-1G A148453-5G
103803-928EA 18 USD
103803-928 103842-040 76719-638 76719-640 77299-654 77299-656
Racecadotril 98%
Racecadotril

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄S
MW: 385.48 g/mol
Boiling Pt: 574.5 °C (760 mm Hg)
Melting Pt: 89 °C
Flash Pt: 301.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00865520
CAS Number: 81110-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR