Rabeprazole sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 2-(((4-(3-methoxypropoxy)-3-methylpyridin-2-yl)methyl)sulphinyl)benzo[d]imidazol-1-ide

A101729-10MG A101729-1MG A101729-100MG A101729-250MG A101729-5G
103796-102EA 74 USD
103796-102 103799-250 76717-570 76717-572 77599-484 77599-486 77666-476
Rabeprazole sodium salt 98%
Rabeprazole sodium salt

Precaución: for Research Use Only

Formula: C₁₈H₂₁N₃O₃S
MW: 381.43 g/mol
Boiling Pt: 603.9 °C (760 mm Hg)
Melting Pt: 150…158 °C
Flash Pt: 319.1 °C
Storage Temperature: Freezer
MDL Number: MFCD02092688
CAS Number: 117976-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR