Quinuclidin-3-yl acetate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Azabicyclo[2.2.2]octan-3-yl acetate

A158629-100MG A158629-10G A158629-250MG A158629-1G
103805-144EA 37 USD
103805-144 103843-360 77190-994 77190-996 77190-998 77319-224
Quinuclidin-3-yl acetate 98%
Quinuclidin-3-yl acetate

Precaución: Sealed in dry, store in freezer, under –20 °C.

Formula: C₉H₁₅NO₂
MW: 169.22 g/mol
Boiling Pt: 221.5 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Freezer
MDL Number: MFCD00468105
CAS Number: 827-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR