Quinoxaline-6-carbaldehyde 97%

Supplier: Ambeed
Warning

A148009-100MG A148009-10G A148009-250MG A148009-1G A148009-5G
103803-866EA 27 USD
103803-866 103841-982 77010-640 77010-642 77010-644 77317-412 77317-414
Quinoxaline-6-carbaldehyde 97%
Quinoxaline-6-carbaldehyde

Precaución: for Research Use Only

Formula: C9H6N2O
MW: 158.1600 g/mol
Boiling Pt: 319.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00805834
CAS Number: 130345-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR