Quinoxaline-2-carboxamide 97%

Supplier: Ambeed
Warning

A263003-100MG A263003-250MG A263003-1G
103812-598EA 169 USD
103812-598 103852-494 76988-222 76988-224 77092-122
Quinoxaline-2-carboxamide 97%
Quinoxaline-2-carboxamide

Precaución: for Research Use Only

Formula: C₉H₇N₃O
Boiling Pt: 430.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01684982
CAS Number: 5182-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR