6-Quinoxalinol 96%

Supplier: Ambeed
Warning

Synonyms: Quinoxalin-6-ol, 6-Hydroxyquinoxaline

A118219-100MG A118219-250MG A118219-5G A118219-1G
103799-008EA 72 USD
103799-008 103824-896 76810-298 76810-300 77297-588 77311-688
6-Quinoxalinol 96%
6-Quinoxalinol

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Boiling Pt: 257.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07364440
CAS Number: 7467-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR