Quinoxalin-5-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxyquinoxaline

A357445-250MG A357445-5G
103826-192EA 21 USD
103826-192 103837-196 76773-316 76840-350
Quinoxalin-5-ol 98%
Quinoxalin-5-ol

Precaución: for Research Use Only

Formula: C8H6N2O
MW: 146.05 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00972004
CAS Number: 17056-99-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR