Quinoxalin-2-amine 97%

Supplier: Ambeed
Warning

A268180-1G A268180-250MG
103812-902EA 78 USD
103812-902 103852-804 77092-552 77262-404
Quinoxalin-2-amine 97%
Quinoxalin-2-amine

Precaución: for Research Use Only

Formula: C₈H₇N₃
Boiling Pt: 297.6 °C (760 mm Hg)
Melting Pt: 155…157 °C
Flash Pt: 159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00094018
CAS Number: 5424-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR