8-Quinolinesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: Quinoline-8-sulphonyl chloride

A455738-1G A455738-5G
103827-846EA 5 USD
103827-846 103838-920 76842-762 77307-394
8-Quinolinesulfonyl chloride 98%
8-Quinolinesulphonyl chloride

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂S
MW: 227.67 g/mol
Boiling Pt: 355.9 °C (760 mm Hg)
Melting Pt: 126…129 °C
Flash Pt: 116.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006808
CAS Number: 18704-37-5
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR