8-Quinolinecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-8-carbaldehyde, 8-Formylquinoline, 8-Quinolinecarboxaldehyde, Quinoline-8-carboxaldehyde

A147064-100G A147064-1G A147064-5G
103803-730EA 13 USD
103803-730 103841-826 76798-466 77317-182 77317-184
8-Quinolinecarbaldehyde 98%
8-Quinolinecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 95…96 °C
Flash Pt: 151.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00805837
CAS Number: 38707-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR