6-Quinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

A104604-25G A104604-1G A104604-5G A104604-10G A104604-500G
103796-742EA 5 USD
103796-742 103822-460 76796-426 77309-324 77309-326 77623-866 77623-868
6-Quinolinecarboxylic acid 98%
6-Quinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 348.7 °C (760 mm Hg)
Melting Pt: 293…294 °C
Flash Pt: 164.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00047613
CAS Number: 10349-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR