5-Quinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

A121026-100MG A121026-10G A121026-250MG A121026-25G A121026-5G A121026-1G A121026-100G
103800-024EA 16 USD
103800-024 76797-346 76797-348 76797-350 76797-352 77297-806 77312-138 77624-004
5-Quinolinecarboxylic acid 98%
5-Quinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 386.5 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 187.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00047608
CAS Number: 7250-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR