3-Quinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

A119268-100G A119268-10G A119268-25G A119268-1G A119268-5G
103799-188EA 5 USD
103799-188 76766-356 76766-358 76766-360 77297-670 77311-860
3-Quinolinecarboxylic acid 98%
3-Quinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 348.7 °C (760 mm Hg)
Melting Pt: 277…280 °C
Flash Pt: 164.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00006770
CAS Number: 6480-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR