3-Quinolinecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde

A111597-100G A111597-10G A111597-250MG A111597-25G A111597-1G A111597-5G
103797-868EA 12 USD
103797-868 103823-686 76765-860 76765-862 76765-864 76765-866 77284-146 77310-500
3-Quinolinecarbaldehyde 98%
3-Quinolinecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 68…71 °C
Flash Pt: 151.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006768
CAS Number: 13669-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR