Quinoline-2-carboximidamide hydrochloride 98%

Supplier: Ambeed
Warning

A120536-250MG A120536-1G A120536-5G
103799-954EA 42 USD
103799-954 76983-226 77312-054 77312-056
Quinoline-2-carboximidamide hydrochloride 98%
Quinoline-2-carboximidamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClN₃
MW: 207.66 g/mol
Melting Pt: 233…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08752296
CAS Number: 110177-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR