8-Quinolineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Quinolin-8-ylboronic acid

A106140-10G A106140-25G A106140-1G A106140-5G
103797-002EA 11 USD
103797-002 103822-756 77270-546 77270-548 77283-448 77309-614
8-Quinolineboronic acid 98%
8-Quinolineboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: >300 °C
Flash Pt: 195.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01114698
CAS Number: 86-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR