Quinolin-6-ylmethanol 97%

Supplier: Ambeed
Warning

A224626-250MG A224626-1G A224626-25G
103819-124EA 31 USD
103819-124 103850-016 76800-934 77088-682 77088-684
Quinolin-6-ylmethanol 97%
Quinolin-6-ylmethanol

Precaución: for Research Use Only

Formula: C10H9NO
MW: 159.1900 g/mol
Boiling Pt: 334.8 °C (760 mm Hg)
Melting Pt: 58…60 °C
Flash Pt: 156.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03789621
CAS Number: 100516-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR