6-Quinolineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Quinolin-6-ylboronic acid, Quinoline-6-boronic acid

A188481-100G A188481-10G A188481-1G A188481-5G A188481-25G A188481-250MG
103816-108EA 17 USD
103816-108 103846-704 76974-756 76974-758 77084-450 77084-452 77302-136 77593-204
6-Quinolineboronic acid 97%
6-Quinolineboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Boiling Pt: 400 °C (760 mm Hg)
Flash Pt: 195.871 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093100
CAS Number: 376581-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR