5-Aminoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: Quinolin-5-amine, Quinolin-5-ylamine

A185417-10G A185417-25G A185417-5G A185417-1G A185417-100G
103815-850EA 9 USD
103815-850 103846-416 76690-696 77084-054 77084-056 77301-972 77625-882
5-Aminoquinoline 97%
5-Aminoquinoline

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 310 °C (760 mm Hg)
Melting Pt: 106…109 °C
Flash Pt: 310 °C
Storage Temperature: Ambient
MDL Number: MFCD00006797
CAS Number: 611-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR