4-Quinolineboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Quinolin-4-ylboronic acid
A251161-25G
A251161-5G
A251161-100MG
A251161-250MG
103811-968EA
42
USD
103811-968
103851-810
77091-186
77091-188
77165-970
77165-972
4-Quinolineboronic acid 98%
4-Quinolineboronic acid
Precaución: for Research Use Only
Formula:
C₉H₈BNO₂ Boiling Pt: 400.3 °C (760 mm Hg) Flash Pt: 195.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD03095174 CAS Number: 371764-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...