3-Quinolineboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Quinoline-3-boronic acid, Quinolin-3-ylboronic acid
A114794-100G
A114794-10G
A114794-25G
A114794-1G
A114794-5G
103798-476EA
9
USD
103798-476
103824-318
77257-854
77257-856
77257-858
77311-144
77311-146
3-Quinolineboronic acid 97%
3-Quinolineboronic acid
Precaución: for Research Use Only
Formula:
C₉H₈BNO₂ Boiling Pt: 400.3 °C (760 mm Hg) Melting Pt: 148…155 °C Flash Pt: 195.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02183527 CAS Number: 191162-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...