(-)-Quinine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
A118859-100G
A118859-10G
A118859-250MG
A118859-25G
A118859-500G
A118859-1G
A118859-5G
A118859-1000G
103799-118EA
9
USD
103799-118
77149-058
77149-060
77149-062
77149-064
77149-066
77297-630
77311-792
77549-384
(-)-Quinine 98%
(-)-Quinine
Precaución: for Research Use Only
Formula:
C₂₀H₂₄N₂O₂ MW: 324.42 g/mol Melting Pt: 173…175 °C Flash Pt: 253.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00198096 CAS Number: 130-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...