Quinazolin-7-amine 98%

Supplier: Ambeed
Warning

A103768-100MG A103768-250MG A103768-1G
103796-608EA 459 USD
103796-608 103822-318 76869-280 76869-282 77309-184
Quinazolin-7-amine 98%
Quinazolin-7-amine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MW: 145.16 g/mol
Boiling Pt: 336 °C (760 mm Hg)
Flash Pt: 183.492 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697884
CAS Number: 101421-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR